Orders LinkContactUsLinkComponent
Druppelflessen, WHEATON®
20 - 410, Colour-21390447|Colour-21390362
  215-4020
Druppelflessen, WHEATON®
  215-4020
Druppelflessen, WHEATON®
Druppelflessen, WHEATON®
20 - 410, Colour-21390447|Colour-21390362
52,00 €
1 * 100 ST
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
  • Schroefdraad:
    20 - 410
  • Seq:
    37
  • Kleur dop:
    Colour-21390447|Colour-21390362