Orders LinkContactUsLinkComponent
Druppelflessen, WHEATON®
Druppelflessen, WHEATON®
13 - 425, wit
Catalogus # 215-4036
Leverancier:   DWK Life Sciences
Druppelflessen, WHEATON®
Catalogus # 215-4036
Leverancier:  DWK Life Sciences
Druppelflessen, WHEATON®
Druppelflessen, WHEATON®
13 - 425, wit
Price
44,60 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Specificaties
  • Schroefdraad:
    13 - 425
  • Seq:
    23
  • Kleur dop:
    wit