Orders LinkContactUsLinkComponent
Filling solutions for combined/reference electrodes
KS100 KCl solution, saturated, 500 ml bottle, 500 mL
Catalogus # HACHC20C300
Leverancier:   Hach
Filling solutions for combined/reference electrodes
Catalogus # HACHC20C300
Leverancier:  Hach
CAS-nummer:  
Filling solutions for combined/reference electrodes
Filling solutions for combined/reference electrodes
KS100 KCl solution, saturated, 500 ml bottle, 500 mL
CAS-nummer:  
Price
59,80 €
Log in voor beschikbaarheid
1 * 500 mL
Product Details & Documents
Filling and storage solutions for combined and reference electrodes.
Specificaties
Specificaties
  • VPE:
    500 mL
  • Omschrijving:
    KS100 KCl solution, saturated, 500 ml bottle