Orders LinkContactUsLinkComponent
Handheld refractometers
Handheld refractometers
0 - 80 °Brix, 0,5 °Brix
Catalogus # 635-0081
Leverancier:   EUROMEX
Handheld refractometers
Catalogus # 635-0081
Leverancier:  EUROMEX
Handheld refractometers
Handheld refractometers
0 - 80 °Brix, 0,5 °Brix
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Single dual or triple scales, analogue, handheld refractometers.
  • Automated temperature compensation (ATC)
  • Calibrated at 20 °C except for RF.5190
  • Requires only a few drops of liquid
Diensten: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Specificaties
  • Meetbereik:
    0 - 80 °Brix
  • Seq:
    07
  • Precisie:
    0,5 °Brix