Hose clips, worm-threaded
20 - 32 mm
Catalogus # BURK8677-2032
Leverancier: BURKLE
Ga naar:
Product Details & Documents
Hose clip sleeve made of stainless steel 1,4016, screw zinc coated, with screw thread.
Documents
BURK8677-0812_TDS_EN BURK8677-1016_TDS_EN BURK8677-1220_TDS_EN
Technische fiche
Specificaties
- Spanwijdte:20 - 32 mm
- Seq:05