Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Spongiosa gouge, -, 5 mm, 250 mm
Catalogus # AESCFL441R
Leverancier:   Aesculap
Gouges, Aesculap®
Catalogus # AESCFL441R
Leverancier:  Aesculap
Gouges, Aesculap®

Spongiosa gouge, -, 5 mm, 250 mm

Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Type:
    -
  • Breedte:
    5 mm
  • Omschrijving:
    Spongiosa gouge
  • Lengte:
    250 mm
  • Seq:
    54
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.