Bone curettes, Aesculap®
Bone curette, type Lempert (Fig. 000), with 2,0 mm end, 215 mm
Catalogus # AESCFK676R
Leverancier: Aesculap
Ga naar:
Product Details & Documents
Specificaties
- Omschrijving:Bone curette, type Lempert (Fig. 000), with 2,0 mm end
- Lengte:215 mm
- Seq:35
- Cat. No.:AESCFK676R