Bone curettes, Aesculap®
Bone curette, type Daubenspeck (Fig. 0), with 5,2 mm end, angled upwards, 200 mm
Catalog # AESCFK665R
Supplier: Aesculap
Jump to:
Product Details & Documents
Specifications
- Omschrijving:Bone curette, type Daubenspeck (Fig. 0), with 5,2 mm end, angled upwards
- Lengte:200 mm
- Seq:33