Bone curettes, Aesculap®
Bone curette, type Daubenspeck (Fig. 00), with 4,4 mm end, angled upwards, 200 mm
Catalogus # AESCFK664R
Leverancier: Aesculap
Ga naar:
Product Details & Documents
Specificaties
- Omschrijving:Bone curette, type Daubenspeck (Fig. 00), with 4,4 mm end, angled upwards
- Lengte:200 mm
- Seq:32