Bone curettes, Aesculap®
Bone curette, Schede type (Fig. 000), with 2,5 mm end, 170 mm
Catalogus # AESCFK621R
Leverancier: Aesculap
Ga naar:
Product Details & Documents
Specificaties
- Omschrijving:Bone curette, Schede type (Fig. 000), with 2,5 mm end
- Lengte:170 mm
- Seq:08