Bone curettes, Aesculap®
Bone curette, Bruns type (Fig. 3), with 10 mm end, 165 mm
Catalogus # AESCFK616R
Leverancier: Aesculap
Ga naar:
Product Details & Documents
Specificaties
- Omschrijving:Bone curette, Bruns type (Fig. 3), with 10 mm end
- Lengte:165 mm
- Seq:06