Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Micro scissors for endoscopic work, Aesculap® MINOP®
Micro scissors for endoscopic work, Aesculap® MINOP®
FF386R, blunt/blunt, 265 mm
Catalogus # AESCFF386R
Leverancier:   Aesculap
Micro scissors for endoscopic work, Aesculap® MINOP®
Catalogus # AESCFF386R
Leverancier:  Aesculap
Micro scissors for endoscopic work, Aesculap® MINOP®
FF386R, blunt/blunt, 265 mm
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Rigid micro scissors for advanced intraventricular Neuroendoscopy.
  • Very delicate
Ø = 2 mm
Bestelinformatie: Complete instrument consists of a handle, an outer tube, and a jaw part with inner tube. Replacements can be ordered separately.
Specificaties
  • Type:
    FF386R, blunt/blunt
  • Lengte:
    265 mm
  • Seq:
    03