Anoscopes, Bensaude, Aesculap®
Anoscope, Bensaude, 55 mm, 23 mm
Catalogus # AESCEA864C
Leverancier: Aesculap
Ga naar:
Product Details & Documents
Anoscopes for abdominal surgery.
Specificaties
- Ø:23 mm
- Omschrijving:Anoscope, Bensaude
- Lengte:55 mm
- Seq:03



