Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Gouge, Lucas, 5 mm, 160 mm
Catalogus # AESCDO717R
Leverancier:   Aesculap
Gouges, Aesculap®
Catalogus # AESCDO717R
Leverancier:  Aesculap
Gouges, Aesculap®

Gouge, Lucas, 5 mm, 160 mm

Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Type:
    Lucas
  • Breedte:
    5 mm
  • Omschrijving:
    Gouge
  • Lengte:
    160 mm
  • Seq:
    22
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.