Dissecting forceps, Aesculap®
Dissecting forceps, Jefferson, Straight, serrated, very fine tips, 180 mm
Catalog # AESCBD226R
Supplier: Aesculap
Jump to:
Product Details & Documents
Stainless steel.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
Delicate dissecting forceps with plateau and many types.
Specifications
- Type:Straight, serrated, very fine tips
- Omschrijving:Dissecting forceps, Jefferson
- Lengte:180 mm
- Seq:14