Pots, laboratory, without lid
Lids for laboratory pots, stainless steel 18/10, for 15,5 pots
Catalogus # BOCH8376
Leverancier: BOCHEM
Ga naar:
Product Details & Documents
Stainless steel 18/10.
- Rounded corners for fast, easy cleaning
- Good chemical resistance
- Very good thermal resistance
Multipurpose pots for a wide range of uses. This series is suitable for induction fields.
Ordering information: Lids to be ordered separately.
Specificaties
- Omschrijving:Lids for laboratory pots, stainless steel 18/10, for 15,5 pots
- Seq:15
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.