Petrischalen
Petri dish, stainless steel, with lid, 100 mm, 15 mm, Niet steriel
Catalog # BOCH8541
Supplier: BOCHEM
Jump to:
Product Details & Documents
Specifications
- Ø:100 mm
- Steriliteit:Niet steriel
- Omschrijving:Petri dish, stainless steel, with lid
- Hoogte:15 mm