Retort rods, with M10 thread
12 mm, 500 mm, M10
Catalogus # BOCH5150
Leverancier: BOCHEM
Ga naar:
Product Details & Documents
Galvanised steel.
- Rods for stand bases
- M10 thread
Specificaties
- Ø:12 mm
- Schroefdraad:M10
- Lengte:500 mm
- Cat. No.:BOCH5150