Orders LinkContactUsLinkComponent
Filtering Bottles, GL 32
Filtering bottle, GL 32, 500 ml
Catalogus # ROBU34500
Leverancier:   ROBU GLASFILTER
Filtering Bottles, GL 32
Catalogus # ROBU34500
Leverancier:  ROBU GLASFILTER
Filtering Bottles, GL 32
Filtering Bottles, GL 32
Filtering bottle, GL 32, 500 ml
Price
90,80 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
ROBU® filtering bottle with GL 32 thread, low pressure resistant with screw-on plastic hose, protective coating, sterilisable. Made up of borosilicate glass 3.3 and plastic.
  • High-quality GL 32 screw cap
  • Includes aperture for easy access to contents
  • Complete set with durable silicone washer for a secure seal
  • Made from chemical-resistant materials
  • Provides a tight and leak-proof fit
  • Ensures optimal protection against contamination
  • Reusable and easy to clean
ROBU caps with aperture, GL 32, are designed for secure sealing and easy access to containers in laboratory applications. Made from high-quality materials, these caps feature a precisely engineered aperture that allows for the insertion of syringes, tubes, or other instruments without compromising the seal.
The GL 32 thread ensures a secure fit on standard laboratory glassware, providing excellent chemical resistance and durability. Ideal for use in sample storage, titration, and other applications where controlled access to the container is required, ROBU Caps with Aperture, GL 32, offer reliable performance in both routine and specialised laboratory tasks.
Specificaties
  • Capaciteit:
    500 ml
  • Omschrijving:
    Filtering bottle, GL 32
  • Seq:
    2