Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Toebehoren
Rack, reinforced, stainless steel, with 1 set of 4 fasteners, max load of 70 kg, FED 400, FP 400, M 400 incubators
Catalogus # BIND8012-0346
Leverancier:   Binder
Toebehoren
Catalogus # BIND8012-0346
Leverancier:  Binder
Toebehoren
Toebehoren
Rack, reinforced, stainless steel, with 1 set of 4 fasteners, max load of 70 kg, FED 400, FP 400, M 400 incubators
Price
503,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Omschrijving:
    Rack, reinforced, stainless steel, with 1 set of 4 fasteners, max load of 70 kg
  • Seq:
    47
  • Voor:
    FED 400, FP 400, M 400 incubators