Passieve dositubes
Product Details & Documents
The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.
- Compact, lightweight, and versatile (can be used anywhere)
- Easy direct measuring and reading
- No sampling pump necessary
The Dositubes use the principle of the natural diffusion of gas.
A Dositube is a glass tube of a specific inner diameter that is filled with a diffuser and chemical reagent in parallel that enable gas to disperse at a constant rate resulting in a longer discolouration and better demarcation. The passive Dositube can then be positioned in the worker's breathing zone where it can be attached with a holder and collar clip.
Spécifications
- Omschrijving:Ammonia (3DL)
- Parameter:Ammonia
- Meetbereik:0,1 - 10 ppm
- Meetduur:1 - 10 h
- Cat. No.:151-0087
Accessories
Informations sur le produit
- N° de catalogueOmschrijvingParameterMeetbereikDisponibilitéPrix
Informations sur le produit
- N° de catalogueOmschrijvingDisponibilitéPrix
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.