Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Instrument trays
Instrument trays
240×355×17 mm
Catalogus # BURK4230-3036
Leverancier:   BURKLE
Instrument trays
Catalogus # BURK4230-3036
Leverancier:  BURKLE
Instrument trays
Instrument trays
240×355×17 mm
Price
31,70 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Melamine, white.
  • Tray without lid
    Suitable for use with foodstuffs
    Not autoclavable
    Not suitable for high temperatures
Bestelinformatie: Lid available as accessory
Specificaties
  • b×d×h:
    240×355×17 mm