Instrument trays
240×355×17 mm
Catalogus # BURK4230-3036
Leverancier: BURKLE
Ga naar:
Product Details & Documents
Melamine, white.
Tray without lid
Suitable for use with foodstuffs
Not autoclavable
Not suitable for high temperatures
Bestelinformatie: Lid available as accessory
Specificaties
- b×d×h:240×355×17 mm