Instrument trays
180×240×17 mm
Catalogus # BURK4230-3024
Leverancier: BURKLE
Ga naar:
Product Details & Documents
Melamine, white.
Tray without lid
Suitable for use with foodstuffs
Not autoclavable
Not suitable for high temperatures
: Lid available as accessory
Specificaties
- b×d×h:180×240×17 mm