Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Xyleen ≥98%, TECHNICAL

Metal drum, 25 L
Catalog # 28973.465
Flammable
Health
Irritant
Supplier:   VWR Chemicals
Xyleen ≥98%, TECHNICAL
Catalog # 28973.465
Supplier:  VWR Chemicals
CAS Number:  1330-20-7
Xyleen ≥98%, TECHNICAL
Xyleen ≥98%, TECHNICAL

Metal drum, 25 L

CAS Number:  1330-20-7
Price
218,00 €
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Dimethylbenzene
  • CAS Number:1330-20-7
  • Formula:C₆H₄(CH₃)₂
  • MDL Number:MFCD00077264
  • Molecular Weight:106.17 g/mol
  • Merck Index:13,10136
  • Boiling Pt:140 °C (1013 hPa)
  • Melting Pt:–34 °C
  • Density:0,86 g/cm³ (20 °C)
  • Flash Pt:25 °C
  • Storage Temperature:Kamertemperatuur
  • EINECS:215-535-7
  • UN:1307
  • ADR:3,III
  • REACH:01-2119488216-32
Specifications

Specification Test Results

  • Assay:
    Min. 98%

Specifications

  • Verpakking:
    Metal drum
  • VPE:
    25 L

Specifications