Volumetric overflow flasks, without stopper
43,5 ml, Overflow flask
Catalogus # NEUB0372-10-0435
Leverancier: NEUBERT VOLUME GLASSWAERE
Volumetric overflow flasks, without stopper
Catalogus # NEUB0372-10-0435
Leverancier: NEUBERT VOLUME GLASSWAERE
Ga naar:
Product Details & Documents
These volumetric overflow flasks are made of borosilicate glass 3.3.
- Calibrated to deliver (TD, Ex)
- Cut and ground exactly at the volume-mark
- Without stopper
Specially designed for water analysis.
Specificaties
- Capaciteit:43,5 ml
- Omschrijving:Overflow flask
- Seq:01