Microplates, 1536-well
µClear-bodem, zwart, gemiddeld bindend, zwart, 1536, -
Catalogus # 738-0089
Leverancier: Greiner Bio-One
Microplates, 1536-well
Catalogus # 738-0089
Leverancier: Greiner Bio-One
Ga naar:
Product Details & Documents
PS, without lid. Ideal for high throughput screening applications, these plates are available as clear bottom variants (in clear PS) and black or white in LUMITRAC™ or FLUOTRAC™ quality.
- HiBase versions designed for top reading instruments, µClear® LoBase plates for bottom reading instruments
- Solid bottom plates available with high binding (MICROLON™ 600, LUMITRAC™ 600 or FLUOTRAC™ 600) or medium binding (LUMITRAC™ 200 or FLUOTRAC™ 200) surfaces
- Rounded square well design combines the advantages of the square well (flexible working volume, 3 to 10 µl) with the advantages of a round well (reduced wicking and bubbling)
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Working volume: 3 - 10 µl
Specificaties
- Aant. wells:1536
- Steriel:-
- Kleur:zwart
- Omschrijving:µClear-bodem, zwart, gemiddeld bindend
- Cat. No.:738-0089
- Milieu certificering:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Lage impact op de fabricage