Microplates, 96-well
Zwart, sterk bindend, steriel, 40 - 340 µl, 96, +, 392 µl
Leverancier: Greiner Bio-One
Ga naar:
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Specificaties
- Aant. wells:96
- Aanbevolen werkvolume:40 - 340 µl
- Steriel:+
- Omschrijving:Zwart, sterk bindend, steriel
- Seq:20
- Totaalvolume:392 µl
- Cat. No.:738-0023
- Milieu certificering:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Lage impact op de fabricage