Order Entry
Belgium
ContactUsLinkComponent
CELLMASTER# roller bottles 1XL, van heldere, breukvaste kunststof (PET) non pyrogenic , DNase/RNase free, oppervl. 1800 cm, vul volume: 4700 ml, graduering 50-1500 ml, lengte 500 mm, diam. 122 mm, per 22 verpakt STERIEL 1 * 22 ST
Catalogus # 736-0234
Leverancier:  Greiner Bio-One
CELLMASTER# roller bottles 1XL, van heldere, breukvaste kunststof (PET) non pyrogenic , DNase/RNase free, oppervl. 1800 cm, vul volume: 4700 ml, graduering 50-1500 ml, lengte 500 mm, diam. 122 mm, per 22 verpakt STERIEL 1 * 22 ST
Catalogus # 736-0234
Leverancier:  Greiner Bio-One
Frequently Bought Together