BUFFER SOLUTION PH 8.5 1 * 500 mL
Catalog # PRLS85199.260
Supplier: PROLABO CMP
Frequently Bought Together
Nitril handschoenen, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Serologische pipetten, standaardreeks
Supplier: VWR Collection
VWR®, Nitrilhandschoenen
Supplier: VWR Collection
VWR®, gezichtstissues
Supplier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH-teststrips, MQuant®, Supelco®
Supplier: Merck
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrilhandschoenen
Supplier: VWR Collection
VWR®, Transferpipetten, Hoge Prestaties
Supplier: VWR Collection