Cryodozen B40/D40
geel, 130×130×45/55 mm
Catalogus # 211-0530
Leverancier: GLW
Ga naar:
Product Details & Documents
Cryoboxes, PP, natural or coloured, without divider.
- Autoclavable up to +121 °C and use in freezer down to –90 °C
- The included clear lid offers a view of the contents without opening the cryobox
- Without drainage holes in base for catching drips or store fluids (leakproof only at bottom up to 40 mm)
- For light sensitive samples please choose the colour Black/black
The boxes comes without compartments, in addition they can be used with GLW dividers for individual configuration.They are suitable to store non standardised tubes, stone samples, small items and more. Inner dimension of the box: 124×124 mm, outer dimension of the box: 130×130 mm.
D40-series
Empty box, without divider. Bottom part of the box – height 45 mm / Lid - height 0 up to 10 mm
D60-series
Empty box, without divider. Bottom part of the box – height 45 mm / Lid - height 7 mm
D70-series
Empty box, without divider. Bottom part of the box – height 45 mm / Lid - height 30 up to 35 mm
D90-series
Empty box, without divider. Bottom part of the box – height 45 mm / Lid - height 45 up to 50 mm.
Documents
GLWAD40_TDS_EN
Datasheet
Specificaties
- Uitw. b×d×h:130×130×45/55 mm
- Kleur:geel
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Etikethouder, transparant, PP, met 20 papieren etiketten, 34 × 100 mmEtikethouder, transparant, PP, met 20 papieren etiketten, 34 × 100 mmGLWAL10Log in voor beschikbaarheid1 * 20 ST - 8,75 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Dividers, PP, natural, length 121 mm, height 27 mmDividers, PP, natural, length 121 mm, height 27 mm211-0349Log in voor beschikbaarheid1 * 1 ST - 0,43 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Dividers, PP, natural, 6×6 compartments 19×19 mmDividers, PP, natural, 6×6 compartments 19×19 mmGLWAZP66Log in voor beschikbaarheid1 * 10 ST - 4,37 €
Productinformatie
- Catalogus #BeschikbaarheidPrice
- 5×5 vakjes van 24×24 mm
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Divider, cardboard, natural, 10×10, compartments size 11,6×11,6 mmDivider, cardboard, natural, 10×10, compartments size 11,6×11,6 mm211-0537Log in voor beschikbaarheid1 * 1 ST - 1,97 €



