Orders LinkContactUsLinkComponent
Accessories for extractors
Accessories for extractors
Extraction chamber B-811, 52 mm, 250 ml
Catalog # BUCH036710
Supplier:   BUCHI
Accessories for extractors
Catalog # BUCH036710
Supplier:  BUCHI
Accessories for extractors
Extraction chamber B-811, 52 mm, 250 ml
Price
1 220,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Omschrijving:
    Extraction chamber B-811, 52 mm, 250 ml
  • Seq:
    04