Instrument boxes
zilver, 90×180×40 mm
Catalogus # AGEN8031
Leverancier: Agencinox
Ga naar:
Product Details & Documents
Aluminium, anodised.
- Smooth surfaces for fast, easy cleaning
- Coloured
- Good chemical resistance, very good thermal resistance
Specificaties
- Kleur:zilver
- Seq:03
- b×d×h:90×180×40 mm