Weegscheppen
Weegschep met knop, 235 mm, 31 mm
N° de catalogue BOCH3602
Fournisseur: BOCHEM
Jump to:
Product Details & Documents
These weighing scoops with knob are made made of 18/10 stainless steel. Available with or without green PTFE coating.
Specifications
- Breedte:31 mm
- Omschrijving:Weegschep met knop
- Lengte:235 mm
- Seq:03