Methyleenchloride stabilised Ph. Eur.
Glass bottle, 1 L
Leverancier: VWR Chemicals
Methyleenchloride stabilised Ph. Eur.
Catalogus # 23365.290
Leverancier: VWR Chemicals
CAS-nummer: 75-09-2
Ga naar:
Product Details & Documents
Synoniemen: Methylene chloride, DCM
- CAS-nummer:75-09-2
- Formule:CH₂Cl₂
- MDL-nummer:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck index:12,06140
- Kokend Pt:39,8 °C (1013 hPa)
- Smelten Pt:–95 °C
- Dichtheid:1,322 g/cm³ (20 °C)
- EINECS:200-838-9
- ADR:6.1,III
- :Stabilised with ethanol 0,1 %
- Temperatuur bij opslag:Kamertemperatuur
- UN:1593
- BEREIKEN:01-2119480404-41
Specificaties
Specificatie testresultaten
- Appearance:Clear colourless liquid
- Identification B:Passes test
- Identification C:Passes test
- Acidity:Passes test
- Relative density:1.320 - 1.332
- Refractive index (20°C):1.423 - 1.425
- Ethanol (V/V):Max. 2 %
- 2-Methyl-2-butene (V/V):Max. 300 ppm
- Total of other impurities (V/V):Max. 0.1 %
- Impurity A (V/V):Max. 10 ppm
- Impurity B (V/V):Max. 50 ppm
- Free chlorine:Passes test
- Heavy metals (as Pb):Max. 1 ppm
- Residue on evaporation:Max. 20 ppm
- Water:Max. 0.02 %
- As (Arsenic):max. 0,1 ppm
- Cd (Cadmium):max. 0,1 ppm
- Co (Cobalt):max. 0,1 ppm
- Other residual solvents:Unlikely by manuf.process
Specificaties
- Verpakking:Glass bottle
- VPE:1 L



