Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Copper(I) bromide ≥97%, TECHNICAL

Copper(I) bromide ≥97%, TECHNICAL

1 kg
  23071.290
 
Copper(I) bromide ≥97%, TECHNICAL
  23071.290
 :  7787-70-4
Copper(I) bromide ≥97%, TECHNICAL
Copper(I) bromide ≥97%, TECHNICAL

1 kg

 :  7787-70-4
 
 
   
1 * 1 kg
 
  •  :7787-70-4
  •  :BrCu
  •  :MFCD00010969
  •  :143.45 g/mol
  •  :13,02689
  •  :1345 °C (1013 hPa)
  •  :504 °C
  •  :4,71 g/cm³ (20 °C)
  •  :Density: 4,71 g/cm³ (20 °C)
  •  :Kamertemperatuur
  •  :232-131-6

Documents

 

 

  • Assay:
    Min. 97 %

 

  • VPE:
    1 kg

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.