Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Ethanol, GPR RECTAPUR®

Ethanol, GPR RECTAPUR®

Metal drum, 200 L
Catalog # 20824.558
Flammable
Irritant
Supplier:   VWR Chemicals
Ethanol, GPR RECTAPUR®
Catalog # 20824.558
Supplier:  VWR Chemicals
CAS Number:  64-17-5
Ethanol, GPR RECTAPUR®
Ethanol, GPR RECTAPUR®

Metal drum, 200 L

CAS Number:  64-17-5
Price
1 850,00 €
Log in for availability
1 * 200 L
Product Details & Documents

Caution: WAARSCHUWING: Deze alcohol is gereguleerd. Bel ons.

Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:C₂H₅OH
  • MDL Number:MFCD00003568
  • Molecular Weight:46.07 g/mol
  • Boiling Pt:78,3 °C (1013 hPa)
  • Melting Pt:–117 °C
  • Density:0,7895 g/cm³ (20 °C)
  • Flash Pt:12 °C (gesloten beker)
  • Storage Temperature:Kamertemperatuur
  • EINECS:200-578-6
  • UN:1170
  • ADR:3,II
  • REACH:01-2119457610-43
Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.8 %
  • Assay (V/V):
    96.0 - 96.3 %
  • Acidity:
    Max. 0.0002 meq/g
  • Aldehydes (as CH3CHO):
    Max. 15 ppm
  • Evaporation residue:
    Max. 20 ppm

Specifications

  • Verpakking:
    Metal drum
  • VPE:
    200 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.