Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Tubidity standards for 2100A
Tubidity standards for 2100A
StabCal® standard calibration kit for model 2100A, 1,0 NTU, Sealed vials, 4× sealed vial, 1 KIT
Catalogus # HACH2659105
Leverancier:   Hach
Tubidity standards for 2100A
Catalogus # HACH2659105
Leverancier:  Hach
CAS-nummer:  
Tubidity standards for 2100A
Tubidity standards for 2100A
StabCal® standard calibration kit for model 2100A, 1,0 NTU, Sealed vials, 4× sealed vial, 1 KIT
CAS-nummer:  
Price
464,00 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
Stablcal Stabilised Formazin Turbidity Standards are true formazin dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazin solutions
  • Based on Formazin Primary Standards
  • Enhanced stability saves time and labour
  • Confidence in results
Documents
Specificaties
Specificaties
  • Verpakking:
    Sealed vials
  • VPE:
    1 KIT
  • Turbiditeit:
    1,0 NTU
  • Omschrijving:
    StabCal® standard calibration kit for model 2100A
  • Info VPE:
    4× sealed vial
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.