Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Sodium chloride standard solution, 10 246 mg/l NaCl, 18 000 µS/cm, 100 mL
Catalogus # HACH2307442
Leverancier:   Hach
Conductivity standards
Catalogus # HACH2307442
Leverancier:  Hach
CAS-nummer:  
Conductivity standards
Conductivity standards
Sodium chloride standard solution, 10 246 mg/l NaCl, 18 000 µS/cm, 100 mL
CAS-nummer:  
Price
31,30 €
Log in voor beschikbaarheid
1 * 100 mL
Product Details & Documents
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Specificaties
Specificaties
  • VPE:
    100 mL
  • Omschrijving:
    Sodium chloride standard solution, 10 246 mg/l NaCl
  • Geleidbaarheid:
    18 000 µS/cm