Orders LinkContactUsLinkComponent
Conductivity standards
Sodium chloride standard solution, 995 mg/l TDS, 1000 mg/l Nacl, 1990 µS/cm, 100 mL
Catalogus # HACH210542
Leverancier:   Hach
Conductivity standards
Catalogus # HACH210542
Leverancier:  Hach
CAS-nummer:  
Conductivity standards
Conductivity standards
Sodium chloride standard solution, 995 mg/l TDS, 1000 mg/l Nacl, 1990 µS/cm, 100 mL
CAS-nummer:  
Price
33,60 €
Log in voor beschikbaarheid
1 * 100 mL
Product Details & Documents
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Specificaties
Specificaties
  • VPE:
    100 mL
  • Omschrijving:
    Sodium chloride standard solution, 995 mg/l TDS, 1000 mg/l Nacl
  • Geleidbaarheid:
    1990 µS/cm
Specificaties