Orders LinkContactUsLinkComponent
Visual comparator test kits, phosphate
Orthophosphate*, 0 - 4,5 mg/l, 100, 1 KIT
Catalog # HACH147500
Supplier:   Hach
Visual comparator test kits, phosphate
Catalog # HACH147500
Supplier:  Hach
CAS Number:  
Visual comparator test kits, phosphate
Visual comparator test kits, phosphate
Orthophosphate*, 0 - 4,5 mg/l, 100, 1 KIT
CAS Number:  
Price
167,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case for 100 tests
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • VPE:
    1 KIT
  • Omschrijving:
    Orthophosphate*
  • Tests per kit:
    100
  • Meetbereik:
    0 - 4,5 mg/l
  • Cat. No.:
    HACH147500
Specifications
Accessories
Product Information
  • Catalog #
    Omschrijving
    VPE
    Availability
    Price
  • HACH11032H
    Ammonium molybdate reagent for phosphate determinations, 100 ml
    1
    Log in for availability
    1 * 1 ST - 27,90 €