Orders LinkContactUsLinkComponent
Empty solid phase extraction cartridges, Bond Elut™
Empty solid phase extraction cartridges, Bond Elut™
Cartridge, 60 ml
Catalog # 551-0361
Supplier:   VARIAN
Empty solid phase extraction cartridges, Bond Elut™
Catalog # 551-0361
Supplier:  VARIAN
Empty solid phase extraction cartridges, Bond Elut™
Empty solid phase extraction cartridges, Bond Elut™
Cartridge, 60 ml
Price
180,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Bond Elut empty SPE cartridges are made with high purity polypropylene for cleaner extracts.
  • Uniform batch-to-batch size for consistent performance
  • Cartridges are available from 1 to 60 ml
Ordering information: Frits has to be ordered separately
Specifications
  • Kolomtype:
    Cartridge
  • Capaciteit:
    60 ml
  • Seq:
    06
Accessories
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • 551-0354
    Adapter caps for 1, 3 and 6 ml Bond Elut™ cartridges
    Log in for availability
    1 * 15 ST - 98,00 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • 551-0355
    Luer stopcocks
    Log in for availability
    1 * 15 ST - 142,00 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • VARI12131035
    Adapter caps for3 ml Gilson ASPEC SPE systems
    Log in for availability
    1 * 1.000 ST - 804,00 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • VARI12131034
    Adapter caps for1ml Gilson ASPEC SPE systems
    Log in for availability
    1 * 1.000 ST - 739,00 €