Cryodozen B72
Product Details & Documents
Cryoboxes made of PP, natural or coloured.
- Autoclavable up to +121°C and use in freezer down to –90 °C
- The included clear lid offers a view of the contents without opening the cryo-box
- Drainage holes in base for easy opening the box at low temperatures
- For light sensitive samples please choose the colour black/black
Designed for 1,5 and 2 ml test tubes, Eppendorf Safe-Lock®, cryotubes, sample vials, microcentrifuge tubes 3810X and flex-tubes generic tube size up to 55 mm height and 12,5 mm diameter. Suitable for 81 samples, arranged in 9×9 compartments with 12,5 mm diameter.
B50-Series:
Designed for 1,5 and 2 ml test tubes, Eppendorf Safe-Lock®, cryotubes, sample vials, microcentrifuge tubes 3810X and flex-tubes. Generic tube size up to 55 mm height and 12,5 mm diameter.
B69-Series:
Designed for 1,5 and 2 ml test tubes, Eppendorf Safe-Lock®, cryotubes, sample vials, microcentrifuge tubes 3810X and flex-tubes. Generic tube size up to 52 mm height and 12,5 mm diameter.
B80-Series:
Designed for 3,6 mm tubes. Generic tube size up to 80 mm height and 12,5 mm diameter.
B99-Series:
Designed for 5 ml tubes. Generic tube size up to 95 mm height and 12,5 mm diameter.
Specificaties
- Compartimenten:9×9
- Uitw. b×d×h:130×130×75/80 mm
- Kleur:geel
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Etikethouder, transparant, PP, met 20 papieren etiketten, 34 × 100 mmEtikethouder, transparant, PP, met 20 papieren etiketten, 34 × 100 mmGLWAL10Log in voor beschikbaarheid1 * 20 ST - 8,75 €



