Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH test kits, model 17

pH test kits, model 17

pH, mid range, model 17F, pH 5,6 - 8,4, Bromothymol blue, 1 KIT
Catalogus # HACH147006
Leverancier:   Hach
pH test kits, model 17
Catalogus # HACH147006
Leverancier:  Hach
CAS-nummer:  
pH test kits, model 17
pH test kits, model 17

pH, mid range, model 17F, pH 5,6 - 8,4, Bromothymol blue, 1 KIT

CAS-nummer:  
Price
183,00 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents

It is a simple, low cost colour disc method which is one of the most accurate visual test kit methods for pH.

Levering: Kit includes indicator reagent, colour disc, comparator box, two viewing tubes, instructions and carrying case.

Specificaties

Specificaties

  • Chemische methode:
    Bromothymol blue
  • VPE:
    1 KIT
  • Omschrijving:
    pH, mid range, model 17F
  • Meetbereik:
    pH 5,6 - 8,4

Specificaties

Accessories

Productinformatie

  • Catalogus #
    VPE
    Beschikbaarheid
    Price
  • Colour viewing tube, glass
    Colour viewing tube, glass
    HACH173006
    6
    Log in voor beschikbaarheid
    1 * 6 ST - 47,80 €

Productinformatie

  • Catalogus #
    VPE
    Beschikbaarheid
    Price
  • Color disc pH, phenol red, 6,6 - 8,4
    Color disc pH, phenol red, 6,6 - 8,4
    HACH9261100
    1
    Log in voor beschikbaarheid
    1 * 1 ST - 109,00 €

Productinformatie

  • Catalogus #
    VPE
    Beschikbaarheid
    Price
  • Stoppers for glass viewing tube
    Stoppers for glass viewing tube
    HACH173106
    6
    Log in voor beschikbaarheid
    1 * 6 ST - 19,50 €

Productinformatie

  • Catalogus #
    VPE
    Beschikbaarheid
    Price
  • Colour comparator box
    Colour comparator box
    HACH173200
    1
    Log in voor beschikbaarheid
    1 * 1 ST - 23,20 €

Productinformatie

  • Catalogus #
    VPE
    Beschikbaarheid
    Price
  • Colour viewing tube, plastic with cap
    Colour viewing tube, plastic with cap
    HACH4660004
    4
    Log in voor beschikbaarheid
    1 * 4 ST - 32,40 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.