Orders LinkContactUsLinkComponent
Microtest manifolds
Microtest manifolds
Microtest manifold, SS/PTFE, bent, 24-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
Catalogus # DRUM3-000-103
Leverancier:   DRUMMOND
Microtest manifolds
Catalogus # DRUM3-000-103
Leverancier:  DRUMMOND
Microtest manifolds
Microtest manifold, SS/PTFE, bent, 24-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
Price
916,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Stainless steel/PTFE manifolds allow multiple test positions to be fed, rinsed, or aspirated quickly and easily.
  • Autoclavable
  • Use for both sample delivery and aspiration operations
  • Supplied complete with cleaning wire for easy cleaning
  • Positive Luer lock connection
Specificaties
  • Omschrijving:
    Microtest manifold, SS/PTFE, bent, 24-place for 384 well plate, 4,5 mm centre-to-centre, pin length 11 mm
  • Seq:
    11