Multiparameter test kits, model HA-62
For hardness (as CaCO₃), iron and pH, HA-62, Hardness: 17,19 - 342,36 mg/l, Iron: 0,1 - 5 mg/l, pH: 6,5 - 8,5, 1 KIT
Catalog # HACH183700
Supplier: Hach
Jump to:
Product Details & Documents
A multi-parameter test kit for hardness, iron and pH, which provides an easy-to-use, cost effective way to determine the quality of water.
- Popular tests in one rugged kit
- Uses color disc and drop count titration methods
Specifications
Specifications
- VPE:1 KIT
- Omschrijving:For hardness (as CaCO₃), iron and pH, HA-62
- Meetbereik:Hardness: 17,19 - 342,36 mg/l, Iron: 0,1 - 5 mg/l, pH: 6,5 - 8,5
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.