Orders LinkContactUsLinkComponent
Comparator kits for water testing
Comparator kits for water testing
Nitrates (low concentration), 0 - 1 mg/l, nitraat (zwak bij C°), 50, 1 KIT
Catalog # 705-0055
Supplier:   Hach
Comparator kits for water testing
Catalog # 705-0055
Supplier:  Hach
CAS Number:  
Comparator kits for water testing
Comparator kits for water testing
Nitrates (low concentration), 0 - 1 mg/l, nitraat (zwak bij C°), 50, 1 KIT
CAS Number:  
Price
183,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • VPE:
    1 KIT
  • Omschrijving:
    Nitrates (low concentration)
  • Parameter:
    nitraat (zwak bij C°)
  • Tests per kit:
    50
  • Meetbereik:
    0 - 1 mg/l
Specifications
Accessories
Product Information
  • Catalog #
    Omschrijving
    VPE
    Availability
    Price
  • HACH11032H
    Ammonium molybdate reagent for phosphate determinations, 100 ml
    1
    Log in for availability
    1 * 1 ST - 27,90 €