Orders LinkContactUsLinkComponent
VWR® Drum Keys
VWR® Drum Keys
Drum keys, plastic, for tightening/opening caps for 10 L (DIN 51) plastic drums
Catalogus # 29548.064
Leverancier:   VWR Collection
VWR® Drum Keys
Catalogus # 29548.064
Leverancier:  VWR Collection
VWR® Drum Keys
VWR® Drum Keys
Drum keys, plastic, for tightening/opening caps for 10 L (DIN 51) plastic drums
Price
8,20 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
A selection of tools to assist in opening IBCs, 200 litre drums, plus bottles and other containers with DIN 32, 45, 54 and 80 caps.
Specificaties
  • Omschrijving:
    Drum keys, plastic, for tightening/opening caps for 10 L (DIN 51) plastic drums