Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Xyleen ≥98%, TECHNICAL

Xyleen ≥98%, TECHNICAL

Glass bottle, 2,5 L
Catalog # 28973.328
Flammable
Health
Irritant
Supplier:   VWR Chemicals
Xyleen ≥98%, TECHNICAL
Catalog # 28973.328
Supplier:  VWR Chemicals
CAS Number:  1330-20-7
Xyleen ≥98%, TECHNICAL
Xyleen ≥98%, TECHNICAL

Glass bottle, 2,5 L

CAS Number:  1330-20-7
Price
43,30 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Dimethylbenzene
  • CAS Number:1330-20-7
  • Formula:C₆H₄(CH₃)₂
  • MDL Number:MFCD00077264
  • Molecular Weight:106.17 g/mol
  • Merck Index:13,10136
  • Boiling Pt:140 °C (1013 hPa)
  • Melting Pt:–34 °C
  • Density:0,86 g/cm³ (20 °C)
  • Flash Pt:25 °C
  • Storage Temperature:Kamertemperatuur
  • EINECS:215-535-7
  • UN:1307
  • ADR:3,III
  • REACH:01-2119488216-32
Specifications

Specification Test Results

  • Assay:
    Min. 98%

Specifications

  • Verpakking:
    Glass bottle
  • VPE:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.