Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Xyleen ≥98%, TECHNICAL

Xyleen ≥98%, TECHNICAL

Glass bottle, 1 L
Catalogus # 28973.294
Ontvlambaar
Gezondheid
Irriterend
Leverancier:   VWR Chemicals
Xyleen ≥98%, TECHNICAL
Catalogus # 28973.294
Leverancier:  VWR Chemicals
CAS-nummer:  1330-20-7
Xyleen ≥98%, TECHNICAL
Xyleen ≥98%, TECHNICAL

Glass bottle, 1 L

CAS-nummer:  1330-20-7
Price
34,80 €
Log in voor beschikbaarheid
1 * 1 L
Product Details & Documents
Synoniemen:  Dimethylbenzene
  • CAS-nummer:1330-20-7
  • Formule:C₆H₄(CH₃)₂
  • MDL-nummer:MFCD00077264
  • Molecular Weight:106.17 g/mol
  • Merck index:13,10136
  • Kokend Pt:140 °C (1013 hPa)
  • Smelten Pt:–34 °C
  • Dichtheid:0,86 g/cm³ (20 °C)
  • Flits Pt:25 °C
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:215-535-7
  • UN:1307
  • ADR:3,III
  • BEREIKEN:01-2119488216-32
Specificaties

Specificatie testresultaten

  • Assay:
    Min. 98%

Specificaties

  • Verpakking:
    Glass bottle
  • VPE:
    1 L

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.