Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Sucrose Ph. Eur.

D-(+)-Sucrose Ph. Eur.

Plastic bucket, 25 kg
Catalog # 27483.465
Supplier:   VWR Chemicals
D-(+)-Sucrose Ph. Eur.
Catalog # 27483.465
Supplier:  VWR Chemicals
CAS Number:  57-50-1
D-(+)-Sucrose Ph. Eur.
D-(+)-Sucrose Ph. Eur.

Plastic bucket, 25 kg

CAS Number:  57-50-1
Price
452,00 €
Log in for availability
1 * 25 kg
Product Details & Documents

Caution: NIET VOOR PARENTERAAL GEBRUIK

Synonyms:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS Number:57-50-1
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,08966
  • Melting Pt:169…170 °C
  • Density:1,29 g/cm³ (20 °C)
  • Storage Temperature:Kamertemperatuur
  • EINECS:200-334-9
Specifications

Specification Test Results

  • Appearance:
    Conforms (see CoA/CoS)
  • Identification (IR):
    Passes test
  • Appearance of solution:
    Passes test
  • Conductivity (20°C; 31,3 %):
    Max. 35 µS/cm
  • Specific optical rotation (26 %; water):
    66.3 - 67 °
  • Colour value:
    Max. 45
  • Reducing sugars:
    Passes test
  • SO3 (as SO2):
    Max. 10 ppm
  • Loss on drying (105°C; 3 h):
    Max. 0.1 %
  • Residual solvents:
    Passes test

Specifications

  • Verpakking:
    Plastic bucket
  • VPE:
    25 kg

Specifications